Journal of immunology (Baltimore, Md. : 1950)
Lipin-1 integrates lipid synthesis with proinflammatory responses during TLR activation in macrophages.
PMID 25252959
Lipin-1 is a Mg(2+)-dependent phosphatidic acid phosphatase involved in the de novo synthesis of phospholipids and triglycerides. Using macrophages from lipin-1-deficient animals and human macrophages deficient in the enzyme, we show in this work that this phosphatase acts as a proinflammatory mediator during TLR signaling and during the development of in vivo inflammatory processes. After TLR4 stimulation lipin-1-deficient macrophages showed a decreased production of diacylglycerol and activation of MAPKs and AP-1. Consequently, the generation of proinflammatory cytokines like IL-6, IL-12, IL-23, or enzymes like inducible NO synthase and cyclooxygenase 2, was reduced. In addition, animals lacking lipin-1 had a faster recovery from endotoxin administration concomitant with a reduced production of harmful molecules in spleen and liver. These findings demonstrate an unanticipated role for lipin-1 as a mediator of macrophage proinflammatory activation and support a critical link between lipid biosynthesis and systemic inflammatory responses.