EMAIL THIS PAGE TO A FRIEND
BMC cancer
ALDH1A1 overexpression is associated with the progression and prognosis in gastric cancer.
PMID 25253129
Abstract
Aldehyde dehydrogenase 1 family member A1 (ALDH1A1) is a cancer stem cell marker, and its expression correlates with prognosis in a number of malignancies. The aim of this study is to determine the relationship of ALDH1A1 expression with clinicopathological parameters and prognosis in gastric cancer. ALDH1A1 and matrix metallopeptidase 9 (MMP-9) was evaluated by immunohistochemistry in 216 gastric carcinoma samples. The association between expression of ALDH1A1 and MMP-9, clinicopathological parameters, and prognosis of gastric cancer was examined. ALDH1A1 protein expression was significantly associated with depth invasion, lymph node metastasis and stage of disease (all P<0.05). Both univariate and multivariate analyses revealed that ALDH1A1 was an independent prognostic factor for both overall survival (OS) and recurrence-free survival (RFS) (both P<0.001). Furthermore, ALDH1A1 overexpression was associated with poor prognosis in patients subgroups stratified by tumor size, depth invasion and lymph node metastasis. Moreover, ALDH1A1 was significantly correlated with MMP-9 among 216 gastric cancer tissues (P<0.001). Patients who had ALDH1A1 overexpression, in which tumor cells displayed high invasiveness, had poor OS and shorter RFS. ALDH1A1 plays an important role in tumor aggressiveness and prognosis, and may act as a promising target for prognostic prediction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405445
Anti-ALDH1A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002123
Anti-ALDH1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M9570 Anti-Matrix Metalloproteinase-9 antibody produced in goat, affinity isolated antibody
HPA001238
Anti-MMP9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
AV33090
Anti-MMP9 antibody produced in rabbit, IgG fraction of antiserum
SAB1403542
Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 1G6, purified immunoglobulin, buffered aqueous solution
SAB5300519 Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 5A11, ascites fluid
SAB1402274
Monoclonal Anti-MMP9 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
AMAB90805
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0539, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90804
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0538, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300247
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5G3, ascites fluid
SAB5300248
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5C3, ascites fluid
AMAB90806
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0542, purified immunoglobulin, buffered aqueous glycerol solution
WH0000216M1
Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 1A2, purified immunoglobulin, buffered aqueous solution