EMAIL THIS PAGE TO A FRIEND
Journal of Virology
Hijacking Dlg1 for oncogenic phosphatidylinositol 3-kinase activation in human epithelial cells is a conserved mechanism of human adenovirus E4-ORF1 proteins.
PMID 25253337