EMAIL THIS PAGE TO A FRIEND
Science translational medicine
Treatment of heterotopic ossification through remote ATP hydrolysis.
PMID 25253675
Abstract
Heterotopic ossification (HO) is the pathologic development of ectopic bone in soft tissues because of a local or systemic inflammatory insult, such as burn injury or trauma. In HO, mesenchymal stem cells (MSCs) are inappropriately activated to undergo osteogenic differentiation. Through the correlation of in vitro assays and in vivo studies (dorsal scald burn with Achilles tenotomy), we have shown that burn injury enhances the osteogenic potential of MSCs and causes ectopic endochondral heterotopic bone formation and functional contractures through bone morphogenetic protein-mediated canonical SMAD signaling. We further demonstrated a prevention strategy for HO through adenosine triphosphate (ATP) hydrolysis at the burn site using apyrase. Burn site apyrase treatment decreased ATP, increased adenosine 3',5'-monophosphate, and decreased phosphorylation of SMAD1/5/8 in MSCs in vitro. This ATP hydrolysis also decreased HO formation and mitigated functional impairment in vivo. Similarly, selective inhibition of SMAD1/5/8 phosphorylation with LDN-193189 decreased HO formation and increased range of motion at the injury site in our burn model in vivo. Our results suggest that burn injury-exacerbated HO formation can be treated through therapeutics that target burn site ATP hydrolysis and modulation of SMAD1/5/8 phosphorylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409631
Anti-BGLAP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4504206
Anti-phospho-Smad1 (pSer187) antibody produced in rabbit, affinity isolated antibody
SAB4503779
Anti-phospho-Smad1 (pSer465) antibody produced in rabbit, affinity isolated antibody
SAB4300250
Anti-phospho-SMAD1 (pSer465) antibody produced in rabbit, affinity isolated antibody
AV36678
Anti-RUNX2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36679
Anti-RUNX2 (AB2) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB1412189
ANTI-RUNX2 antibody produced in mouse, clone 6E1, purified immunoglobulin, buffered aqueous solution
SAB1412247
ANTI-RUNX2 antibody produced in mouse, clone 4E5, ascites fluid
SAB1412248
ANTI-RUNX2 antibody produced in mouse, clone 4D7, purified immunoglobulin, buffered aqueous solution
SAB1412249
ANTI-RUNX2 antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1412665
ANTI-RUNX2 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB1412666
ANTI-RUNX2 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1412667
ANTI-RUNX2 antibody produced in mouse, clone 3F5, purified immunoglobulin, buffered aqueous solution
SAB1412668
ANTI-RUNX2 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
HPA022040
Anti-RUNX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106220
Anti-RUNX2 antibody produced in rabbit, affinity isolated antibody
SAB2701834
Anti-smad5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV32006
Anti-SMAD5 antibody produced in rabbit, affinity isolated antibody
SAB4200406
Anti-SMAD5 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA058931
Anti-SMAD5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1412499
ANTI-SOX9 antibody produced in mouse, clone 2A2, purified immunoglobulin, buffered aqueous solution
HPA001758
Anti-SOX9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37986
Anti-SOX9 antibody produced in rabbit, affinity isolated antibody
AV100797
Anti-SOX9 antibody produced in rabbit, IgG fraction of antiserum
SAB4502834
Anti-SOX9 antibody produced in rabbit, affinity isolated antibody
WH0000632M1
Monoclonal Anti-BGLAP antibody produced in mouse, clone 2D4, purified immunoglobulin, buffered aqueous solution
SAB1401030
Monoclonal Anti-BGLAP antibody produced in mouse, clone 2D5, purified immunoglobulin, buffered aqueous solution
WH0000860M1
Monoclonal Anti-RUNX2 antibody produced in mouse, clone 1D8, purified immunoglobulin, buffered aqueous solution
AMAB90591
Monoclonal Anti-RUNX2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0232, purified immunoglobulin, buffered aqueous glycerol solution
SAB1403638
Monoclonal Anti-RUNX2 antibody produced in mouse, clone 4D5, purified immunoglobulin, buffered aqueous solution