PloS one
Elevation of extracellular Ca2+ induces store-operated calcium entry via calcium-sensing receptors: a pathway contributes to the proliferation of osteoblasts.
PMID 25254954
Abstract
The local concentration of extracellular Ca(2+) ([Ca(2+)]o) in bone microenvironment is accumulated during bone remodeling. In the present study we investigated whether elevating [Ca(2+)]o induced store-operated calcium entry (SOCE) in primary rat calvarial osteoblasts and further examined the contribution of elevating [Ca(2+)]o to osteoblastic proliferation. Cytosolic Ca(2+) concentration ([Ca(2+)]c) of primary cultured rat osteoblasts was detected by fluorescence imaging using calcium-sensitive probe fura-2/AM. Osteoblastic proliferation was estimated by cell counting, MTS assay and ATP assay. Agonists and antagonists of calcium-sensing receptors (CaSR) as well as inhibitors of phospholipase C (PLC), SOCE and voltage-gated calcium (Cav) channels were applied to study the mechanism in detail. Our data showed that elevating [Ca(2+)]o evoked a sustained increase of [Ca(2+)]c in a dose-dependent manner. This [Ca(2+)]c increase was blocked by TMB-8 (Ca(2+) release inhibitor), 2-APB and BTP-2 (both SOCE blockers), respectively, whereas not affected by Cav channels blockers nifedipine and verapamil. Furthermore, NPS2143 (a CaSR antagonist) or U73122 (a PLC inhibitor) strongly reduced the [Ca(2+)]o-induced [Ca(2+)]c increase. The similar responses were observed when cells were stimulated with CaSR agonist spermine. These data indicated that elevating [Ca(2+)]o resulted in SOCE depending on the activation of CaSR and PLC in osteoblasts. In addition, high [Ca(2+)]o significantly promoted osteoblastic proliferation, which was notably reversed by BAPTA-AM (an intracellular calcium chelator), 2-APB, BTP-2, TMB-8, NPS2143 and U73122, respectively, but not affected by Cav channels antagonists. Elevating [Ca(2+)]o induced SOCE by triggering the activation of CaSR and PLC. This process was involved in osteoblastic proliferation induced by high level of extracellular Ca(2+) concentration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16609
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
C36H44N2O18
D9754
2-Aminoethyl diphenylborinate, 97%
C14H16BNO
42810
2-Aminoethyl diphenylborinate, for TLC derivatization, ≥97.0%
C14H16BNO
A1076
BAPTA-AM, ≥95% (HPLC)
C34H40N2O18
02672
Calcium atomic spectroscopy standard concentrate 1.00 g Ca, 1.00 g/L, for 1L standard solution, analytical standard
CaCl2
21059
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
CaCl2
94563
Dimethyl sulfoxide, analytical standard
C2H6OS
01934
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
C2H6OS
1211006
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
C2H6OS
D2050000
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
C2H6OS
E0396
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
C14H24N2O10
E3889
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
C14H24N2O10
E4378
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
C14H24N2O10
03777
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
C14H24N2O10
N7634
Nifedipine, ≥98% (HPLC), powder
C17H18N2O6
N114
Nifedipine
C17H18N2O6
N0750000
Nifedipine, European Pharmacopoeia (EP) Reference Standard
C17H18N2O6
1463508
Nifedipine, United States Pharmacopeia (USP) Reference Standard
C17H18N2O6
PHR1290
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
C17H18N2O6
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
S4264
Spermine, BioReagent, amorphous semi-solid, suitable for cell culture
C10H26N4
S3256
Spermine, ≥97%
C10H26N4
85590
Spermine, ≥99.0% (GC)
C10H26N4
55513
Spermine, analytical standard
C10H26N4