EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Separation and determination of impurities in paracetamol, codeine and pitophenone in the presence of fenpiverinium in combined suppository dosage form.
PMID 25255449