EMAIL THIS PAGE TO A FRIEND
The Journal of Pharmacy and Pharmacology
Adrenaline (epinephrine) microcrystal sublingual tablet formulation: enhanced absorption in a preclinical model.
PMID 25256073
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000882
Adrenaline, European Pharmacopoeia (EP) Reference Standard
C9H13NO3