EMAIL THIS PAGE TO A FRIEND
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.]
Clinical correlations with (18)FDG PET scan patterns in solid pseudopapillary tumors of the pancreas: still a surgical enigma?
PMID 25256437
Abstract
There are limited numbers of PET studies of solid pseudopapillary tumors (SPT) of the pancreas. We reviewed the medical records of 37 patients who underwent resection of pancreatic SPT and had been preoperatively evaluated by (18)F-FDG PET or PET/CT scan. Immunohistochemical analysis of glucose transporter-1 (GLUT-1) and hexokinase II (HK-II) was performed. SPT could be categorized into five types according to the morphologic characteristics observed in PET images. Type I (hot FDG uptake in the entire tumor portion) was the most frequent (13, 34.2%), followed by type IV (focal uptake, 12, 31.6%), II (focal defect, 8, 21.1%), III (multiple and geographic uptake, 3, 7.9%), and V (total defective type, 1, 2.6%). The SUVmax in the solid portion of the SPT was 5.3 ± 4.1. The clinical pattern of FDG uptake in SPT was not associated with histopathologic features suggesting malignant potential. The SUVmax of SPT followed a pattern according to pattern of FDG uptake (R(2) = 0.203, p = 0.055), and was significantly associated with adjusted tumor volume (p = 0.001). GLUT-1 was not expressed in SPT, and only eight patients (12.3%) showed mild to moderate expression of HK-II, which was associated with the clinical pattern of SPT in PET images (p < 0.05). SPT of the pancreas could be categorized according to the morphologic patterns observed in PET images. The clinical significance of FDG uptake, glucose metabolism, and clinical usefulness of PET scan in SPT need to be further investigated, and thus this tumor remains a surgical enigma.