EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
In vitro antioxidant and anti-inflammatory activities of Radix Isatidis extract and bioaccessibility of six bioactive compounds after simulated gastro-intestinal digestion.
PMID 25256688