EMAIL THIS PAGE TO A FRIEND
Diabetologia
Vascular heterogeneity between native rat pancreatic islets is responsible for differences in survival and revascularisation post transplantation.
PMID 25257098