EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Concluding the amyloid formation pathway of a coiled-coil-based peptide from the size of the critical nucleus.
PMID 25257178
Abstract
The size of the critical nucleus acting as intermediate in the amyloid formation of a model peptide is calculated. The theoretical approach is based on experimentally determined amyloid formation rates and gives new insights into the amyloid formation pathway.