EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Effects of intestinal microbiota on the bioavailability of geniposide in rats.
PMID 25257195
Abstract
This study investigated the effects of intestinal microbiota on the metabolism of geniposide by using a rat model treated with a mixture of antibiotics. The plasma concentration of geniposide was determined after oral administration in control and antibiotics-treated rats by using liquid chromatography-tandem mass spectrometry. The maximum plasma concentrations (Cmax) of geniposide in control and antibiotics-treated rats were 0.91 ± 0.26 and 1.01 ± 0.04 μg/mL, respectively, and the area under the curve (AUC) values were 7.34 ± 3.32 and 11.9 ± 2.1 μg·h/mL (p < 0.05), respectively. The levels of geniposide in rat feces were 0.64 and 15.6 mg, respectively, in the control and antibiotics-treated groups. Thus, the systemic exposure of geniposide was greater in the antibiotics-treated rats. This may be due to the antibiotic-induced suppression of the metabolic activities of the intestinal microbiota. These results suggest that the gut microbiota may have an impact on the bioavailability of geniposide.