EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Abnormal response of the proliferation and differentiation of growth plate chondrocytes to melatonin in adolescent idiopathic scoliosis.
PMID 25257530
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1380105
Melatonin, United States Pharmacopeia (USP) Reference Standard
C13H16N2O2