EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Gastrointestinal and Liver Physiology
L-Ornithine phenylacetate reduces ammonia in pigs with acute liver failure through phenylacetylglycine formation: a novel ammonia-lowering pathway.
PMID 25258408
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0325000
Alanine, European Pharmacopoeia (EP) Reference Standard
C3H7NO2
368407
Deuterium, 99.96 atom % D
D2
617474
Deuterium, 99.9 atom % D
D2
361860
Deuterium, 99.8 atom % D
D2
488690
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
HD
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
58958
Ethyl acetate, analytical standard
C4H8O2
17023
Ethyl acetate, tested according to Ph.Eur.
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
09-0703
Ethyl acetate, for residue analysis, JIS 5000
C4H8O2
09-0730
Ethyl acetate, for HPLC
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
PHR1481
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
C4H8O2
76524
Glycine, certified reference material, TraceCERT®
C2H5NO2
50058
Glycine, tested according to Ph.Eur.
C2H5NO2
PHR1111
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
C2H5NO2
1295800
Glycine, United States Pharmacopeia (USP) Reference Standard
C2H5NO2
94119
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
C2H5NO2
G0450000
Glycine, European Pharmacopoeia (EP) Reference Standard
C2H5NO2
112003
Hippuric acid, 98%
C9H9NO3
S0450000
Serine, European Pharmacopoeia (EP) Reference Standard
C3H7NO3
306975
tert-Butyl methyl ether, anhydrous, 99.8%
C5H12O
675407
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C5H12O
34875
tert-Butyl methyl ether, for HPLC, ≥99.8%
C5H12O
08603
tert-Butyl methyl ether, analytical standard
C5H12O
20256
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
C5H12O
293210
tert-Butyl methyl ether, HPLC grade, for HPLC, 99.8%
C5H12O