EMAIL THIS PAGE TO A FRIEND
Nanoscale Research Letters
Green synthesis and synergistic catalytic effect ofAg/reduced graphene oxide nanocomposite.
PMID 25258607
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1048
4-Nitrophenol, spectrophotometric grade
C6H5NO3
35836
4-Nitrophenol, PESTANAL®, analytical standard
C6H5NO3
241326
4-Nitrophenol, ReagentPlus®, ≥99%
C6H5NO3
48549
4-Nitrophenol
C6H5NO3
PHR1150
4-NITROPHENOL (Acetaminophen Related Compound F)(Paracetamol Impurity F), Pharmaceutical Secondary Standard; Certified Reference Material
C6H5NO3
N7660
4-Nitrophenol solution, 10 mM
C6H5NO3
40056
4-Nitrophenol solution, certified reference material, 5000 μg/mL in methanol
C6H5NO3
43575
4-Nitrophenol solution, stock solution, for mass spectrometry
C6H5NO3
49053
4-Nitrophenol solution, working solution, for mass spectrometry
C6H5NO3
71320
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
H4BNa
452890
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
H4BNa
480886
Sodium borohydride, granular, 99.99% trace metals basis
H4BNa
452874
Sodium borohydride, granular, 10-40 mesh, 98%
H4BNa
452882
Sodium borohydride, powder, ≥98.0%
H4BNa
247677
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
H4BNa
71318
Sodium borohydride, purum, ≥97.0% (gas-volumetric), tablet
H4BNa
213462
Sodium borohydride, ReagentPlus®, 99%
H4BNa
686018
Sodium borohydride, hydrogen-storage grade, 98%
H4BNa
200972
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
H4BNa
452904
Sodium borohydride solution, SWS, ~12 wt. % in 14 M NaOH
H4BNa
215511
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
H4BNa
S5506
Sodium nitrate, ReagentPlus®, ≥99.0%
NNaO3
S5022
Sodium nitrate, ≥99.0%, plant cell culture tested
NNaO3
229938
Sodium nitrate, 99.995% trace metals basis
NNaO3
15736
Sodium nitrate, analytical standard
NNaO3
S8170
Sodium nitrate, BioXtra, ≥99.0%
NNaO3
221341
Sodium nitrate, ACS reagent, ≥99.0%
NNaO3
71755
Sodium nitrate, BioUltra, ≥99.0% (T)
NNaO3
13444
Sodium nitrate, meets analytical specification of FCC, E251, 99.0-100.5%
NNaO3
28-3440
Sodium nitrate, JIS special grade, ≥99.0%
NNaO3