EMAIL THIS PAGE TO A FRIEND
Journal of Analytical Methods in Chemistry
Extraction and Bioactivity Analysis of Major Flavones Compounds from Scutellaria baicalensis Using In Vitro Assay and Online Screening HPLC-ABTS System.
PMID 25258697