EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Analysis of 21 progestagens in various matrices by ultra-high-performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) with diverse sample pretreatment.
PMID 25260405