EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Modern analytics for naturally derived complex drug substances: NMR and MS tests for protamine sulfate from chum salmon.
PMID 25260409