EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Hydroxycinnamic acids used as external acceptors of electrons: an energetic advantage for strictly heterofermentative lactic acid bacteria.
PMID 25261518