EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
CCN family protein 2 (CCN2) promotes the early differentiation, but inhibits the terminal differentiation of skeletal myoblasts.
PMID 25261584
Abstract
Many studies have reported that CCN family protein 2 (also known as connective tissue growth factor) induces fibrotic response in skeletal muscle, thus emphasizing the pathological role of CCN2 in muscle tissues. However, the physiological role of CCN2 in myogenesis is still unknown. This study clarified the CCN2 functions during myogenesis. Recombinant CCN2 (rCCN2) promoted proliferation and MyoD production in C2C12 cells and primary myoblasts, but inhibited myogenin production. In accordance with these findings, the gene expression levels of myosin heavy chain, which is a marker of terminally differentiated myoblasts and desmin, which is the main intermediate filament protein of muscle cells, were decreased by rCCN2 treatment. In vivo analyses with Ccn2-deficient skeletal muscle revealed decreased proliferating cell nuclear antigen (PCNA)/MyoD double positive cells and muscle hypoplasia. Consistent with this finding, myogenic marker genes and myotube formation were repressed in Ccn2-deficient myoblasts. The protein production of CCN2 was increased in C2C12 myoblasts treated with tumor necrosis factor-α, which is a pro-inflammatory cytokine, suggesting its role in muscle regeneration after inflammation. These findings indicate that CCN2 promotes proliferation and early differentiation but inhibits the terminal differentiation of myoblasts, thus suggesting that CCN2 plays a physiological role in myogenesis.