EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Evaluation of enantiomeric purity of magnesium-L-aspartate dihydrate.
PMID 25262412
Abstract
Magnesium supplementation in form of organic magnesium salts is a very popular practice. We examined the enantiomeric purity of "Magnesium aspartate dihydrate" monographed in the European Pharmacopeia. A chiral capillary zone electrophoresis using (2-hydroxypropyl)-β-cyclodextrin coupled to laser induced fluorescence detection and a HPLC-fluorescence method with chiral derivatization using o-phthaldialdehyde and N-acetyl-L-cysteine as an orthogonal method were developed and validated. Two batch samples of this substance and three drug products containing the salt were analyzed by means of both methods. The concentration of the D-enantiomer of aspartic acid ranged from 0.03 to 0.12%. Simulations of the synthesis revealed that the d-aspartic acid content is elevated if the dissolution of L-aspartic acid was performed at acidic pH values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5474
L-Aspartic acid, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C4H7NO4
A9256
L-Aspartic acid, reagent grade, ≥98% (HPLC)
C4H7NO4
A8949
L-Aspartic acid, BioXtra, ≥99% (HPLC)
C4H7NO4
A7219
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C4H7NO4
11189
L-Aspartic acid, BioUltra, ≥99.5% (T)
C4H7NO4
PHR1104
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C4H7NO4
W365601
L-Aspartic acid, ≥98%, FG
C4H7NO4
51572
L-Aspartic acid, certified reference material, TraceCERT®
C4H7NO4
G5667
L-Glutamic acid, PharmaGrade, Ajinomoto, EP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C5H9NO4
W328502
L-Glutamic acid, 99%, FCC
C5H9NO4
G8415
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
C5H9NO4
49449
L-Glutamic acid, BioUltra, ≥99.5% (NT)
C5H9NO4
95436
L-Glutamic acid, certified reference material, TraceCERT®
C5H9NO4
G1251
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
C5H9NO4
09581
L-Glutamic acid, tested according to Ph.Eur.
C5H9NO4
PHR1107
L-Glutamic acid
C5H9NO4
31458
Magnesia grooves
MgO
31409
Magnesia grooves
MgO
M0088000 Magnesium aspartate dihydrate, European Pharmacopoeia (EP) Reference Standard
C8H12MgN2O8 · 2H2O
M7179
Magnesium carbonate, meets USP testing specifications
CMgO3 · xH2O
PHR1328
Magnesium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
CMgO3 · xH2O
M5421
Magnesium hydroxide, BioXtra, ≥95%
H2MgO2
63081
Magnesium hydroxide, BioUltra, ≥99.0% (KT)
H2MgO2
310093
Magnesium hydroxide, reagent grade, 95%
H2MgO2
632309
Magnesium hydroxide, nanopowder, <100 nm particle size (laser PSA), 99.8% trace metals basis
H2MgO2
529699
Magnesium oxide, ≥99.99% trace metals basis
MgO
203718
Magnesium oxide, 99.99% trace metals basis
MgO
342793
Magnesium oxide, ≥99% trace metals basis, -325 mesh
MgO
220361
Magnesium oxide, -10-+50 mesh, 98%
MgO
342777
Magnesium oxide, fused, chips, -4 mesh, 99.9% trace metals basis
MgO