EMAIL THIS PAGE TO A FRIEND
European Journal of Immunology
Cross-reactivity of hepatitis C virus specific vaccine-induced T cells at immunodominant epitopes.
PMID 25263407
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757349
Fluoroethylene carbonate, 99%
C3H3FO3