EMAIL THIS PAGE TO A FRIEND
American Journal of Hematology
Enrichment of reticulocytes from whole blood using aqueous multiphase systems of polymers.
PMID 25263455
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0731000 Dextran, European Pharmacopoeia (EP) Reference Standard
D0731005 Dextran 1, European Pharmacopoeia (EP) Reference Standard
D0734000 Dextran 10 for calibration, European Pharmacopoeia (EP) Reference Standard
D0737000 Dextran 250 for calibration, European Pharmacopoeia (EP) Reference Standard
D0733000 Dextran 4 for calibration, European Pharmacopoeia (EP) Reference Standard
D0735000 Dextran 40 for calibration, European Pharmacopoeia (EP) Reference Standard
D0738000 Dextran 40 for performance test, European Pharmacopoeia (EP) Reference Standard
D0739000 Dextran 60/70 for performance test, European Pharmacopoeia (EP) Reference Standard
D0736000 Dextran 70 for calibration, European Pharmacopoeia (EP) Reference Standard
D0732000 Dextran Vo, European Pharmacopoeia (EP) Reference Standard
Y0000441
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
C5H4N4O
77662
Hypoxanthine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C5H4N4O
H9377
Hypoxanthine, ≥99.0%
C5H4N4O
H9636
Hypoxanthine, powder, BioReagent, suitable for cell culture
C5H4N4O
M9140
Methylene blue, certified by the Biological Stain Commission
C16H18ClN3S · xH2O
03978
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
C16H18ClN3S
1808
Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
C16H18ClN3S
319112
Methylene Blue solution, 0.05 wt. % in H2O
C16H18ClN3S
77515
Methylene Blue solution, for microscopy
C16H18ClN3S
50484
Methylene Blue solution, for microbiology
C16H18ClN3S
66725
Methylene Blue solution alkaline according to Loeffler, according to Loeffler
C16H18ClN3S
Y0001550
Poly(vinyl alcohol), European Pharmacopoeia (EP) Reference Standard