EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Combinatorial measurement of CDKN1A/p21 and KIF20A expression for discrimination of DNA damage-induced clastogenicity.
PMID 25264741