EMAIL THIS PAGE TO A FRIEND
Experimental Cell Research
Studying p53 family proteins in yeast: induction of autophagic cell death and modulation by interactors and small molecules.
PMID 25265062
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0580
Nutlin-3a, ≥98% (HPLC)
C30H30Cl2N4O4