EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Reply to "Mercury Bioaccumulation and Bioaccumulation Factors for Everglades Mosquitofish as Related to Sulfate: A Re-Analysis of Julian II (2013)".
PMID 25266044