EMAIL THIS PAGE TO A FRIEND
Parasitology research
Propolis enhances the effectiveness of praziquantel in experimental schistosomiasis: biochemical and histopathological study.
PMID 25266217
Abstract
Despite the wide current use of praziquantel (PZQ) in treatment of schistosomiasis, low cure rates have been recorded in many studies. The aim of this study was directed to evaluate the curative effect of propolis (Pps) alone or in combination with PZQ on biochemical, immunological, parasitological, and histological changes associated with experimental schistosomiasis in mice. Schistosoma mansoni-infected mice were divided into two experimental sets, each with four subgroups: (i) untreated, (ii) treated with Pps/day p.o for 4xa0weeks, (iii) treated with PZQ p.o 2 × 500xa0mg/kg bd wt, and (iv) treated with Pps + PZQ as in group ii and iii; all treatments started on the 8th week postinfection, in addition to uninfected group as control for the previous groups. Treatment of infected mice with Pps, although failed to eradicate the worm, significantly reduced the hepatic granuloma number, their lymphocytic infiltration and aggregation, hepatic and splenic myeloperoxidase (MPO) activity and plasma, and liver and thymus nitric oxide (NOx) levels together with normalization of plasma proteins and alleviation of oxidative stress in the examined tissues as evidenced by reduction of malondialdehyde (MDA) and normalization of glutathione (GSH). Promising results were obtained when Pps was given in combination with PZQ, where the anti-schistosomal activity of PZQ was markedly potentiated with complete alleviation and amelioration of the histological and biochemical alteration associated with schistosomiasis. This study highlights the potential usefulness of Pps as an adjunct to PZQ in schistosomiasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3682 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
A9309 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701023 Anti-Mouse IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
A3673 Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701037 Anti-Mouse IgG (γ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701131 Anti-Mouse IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701159 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701165 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701116 Anti-Mouse IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3701105
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3701083
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701095
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
A8924 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A5278 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
A5906 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
A6782 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB3701015 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701004 Anti-Mouse IgG F(ab′)2, highly cross adsorbed-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700993 Anti-Mouse IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701003 Anti-Mouse IgG F(ab′)2-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder