EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Structures of bacterial polynucleotide kinase in a michaelis complex with nucleoside triphosphate (NTP)-Mg2+ and 5'-OH RNA and a mixed substrate-product complex with NTP-Mg2+ and a 5'-phosphorylated oligonucleotide.
PMID 25266383
Abstract
Clostridium thermocellum polynucleotide kinase (CthPnk), the 5'-end-healing module of a bacterial RNA repair system, catalyzes reversible phosphoryl transfer from a nucleoside triphosphate (NTP) donor to a 5'-OH polynucleotide acceptor, either DNA or RNA. Here we report the 1.5-Å crystal structure of CthPnk-D38N in a Michaelis complex with GTP-Mg(2+) and a 5'-OH RNA oligonucleotide. The RNA-binding mode of CthPnk is different from that of the metazoan RNA kinase Clp1. CthPnk makes hydrogen bonds to the ribose 2'-hydroxyls of the 5' terminal nucleoside, via Gln51, and the penultimate nucleoside, via Gln83. The 5'-terminal nucleobase is sandwiched by Gln51 and Val129. Mutating Gln51 or Val129 to alanine reduced kinase specific activity 3-fold. Ser37 and Thr80 donate functionally redundant hydrogen bonds to the terminal phosphodiester; a S37A-T80A double mutation reduced kinase activity 50-fold. Crystallization of catalytically active CthPnk with GTP-Mg(2+) and a 5'-OH DNA yielded a mixed substrate-product complex with GTP-Mg(2+) and 5'-PO4 DNA, wherein the product 5' phosphate group is displaced by the NTP γ phosphate and the local architecture of the acceptor site is perturbed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254118
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Mg
474754
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Mg
465992
Magnesium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Mg
254126
Magnesium, 20-230 mesh, reagent grade, 98%
Mg
63040
Magnesium, grit, ≥99.0% (KT)
Mg
253987
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Mg
465666
Magnesium, ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis
Mg
13112
Magnesium, powder, ≥99%
Mg
266302
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Mg
13103
Magnesium, ribbon, ≥99.5% Mg basis
Mg
299405
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Mg
403148
Magnesium, turnings, ~1/8 in., 99.95% trace metals basis
Mg
200905
Magnesium, turnings, reagent grade, 98%
Mg
63035
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Mg
723312
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Mg
723320
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Mg
735779
Magnesium, ingot, ≥99% trace metals basis
Mg
734365
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Mg
735760
Magnesium, plate, 99.95%
Mg
GF86050199
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Mg
GF67975172
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF80461964
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF57746166
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Mg
GF18270637
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Mg
GF96495609
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Mg
GF04987547
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Mg
GF23637371
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Mg
GF83427745
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Mg
GF86178189
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Mg
GF83829178
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Mg