EMAIL THIS PAGE TO A FRIEND
Journal of Hazardous Materials
Pharmaceuticals and pesticides in reclaimed water: Efficiency assessment of a microfiltration-reverse osmosis (MF-RO) pilot plant.
PMID 25269743