EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Medicine
Tanshinone IIA inhibits human gastric carcinoma AGS cell growth by decreasing BiP, TCTP, Mcl?1 and Bcl?xL and increasing Bax and CHOP protein expression.
PMID 25270224
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1351
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6