EMAIL THIS PAGE TO A FRIEND
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA
Increased serum fibroblast growth factor-23 and decreased bone turnover in patients with systemic lupus erythematosus under treatment with cyclosporine and steroid but not steroid only.
PMID 25270396
Abstract
In patients with systemic lupus erythematosus (SLE), low bone mineral density (BMD) is associated with increased age, prolonged disease, low body mass index (BMI), and overlap with rheumatoid arthritis (RA). Elevated fibroblast growth factor (FGF)-23 in cyclosporine A (CsA) users with SLE are associated with decreased active vitamin D and osteocalcin. The objective of this study was to investigate the steroid and CsA effect on bone metabolism and serum FGF-23 in SLE patients. Seventy-two SLE patients and 10 age- and sex-matched healthy individuals underwent blood tests for bone metabolic biomarkers and FGF-23, and lumbar spine dual-energy X-ray absorptiometry for BMD. Comparisons between patients and controls were made in premenopausal women/men younger than 50 years and postmenopausal women/men older than 50 years separately. SLE patients had more frequent low Z-score (≤-2.0, 8.5 vs. 0%), osteopenia (-2.5