EMAIL THIS PAGE TO A FRIEND
Psychopharmacology Series
Role of ionotropic glutamate receptors in delay and probability discounting in the rat.
PMID 25270726
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M107
(+)-MK-801 hydrogen maleate, powder
C16H15N · C4H4O4