EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Tri- and tetra-nuclear polypyridyl ruthenium(II) complexes as antimicrobial agents.
PMID 25271478
Abstract
A series of inert tri- and tetra-nuclear polypyridylruthenium(II) complexes that are linked by the bis[4(4'-methyl-2,2'-bipyridyl)]-1,n-alkane ligand ("bb(n)" for n = 10, 12 and 16) have been synthesised and their potential as antimicrobial agents examined. Due to the modular nature of the synthesis of the oligonuclear complexes, it was possible to make both linear and non-linear tetranuclear ruthenium species. The minimum inhibitory concentrations (MIC) of the ruthenium(II) complexes were determined against four strains of bacteria--Gram positive Staphylococcus aureus (S. aureus) and methicillin-resistant S. aureus (MRSA), and Gram negative Escherichia coli (E. coli) and Pseudomonas aeruginosa (P. aeruginosa). In order to gain an understanding of the relative antimicrobial activities, the cellular uptake and water-octanol partition coefficients (log P) were determined for a selection of the ruthenium complexes. Although the trinuclear complexes were the most lipophilic based upon log P values and showed the greatest cellular uptake, the linear tetranuclear complexes were generally more active, with MIC values <1 μM against the Gram positive bacteria. Similarly, although the non-linear tetranuclear complexes were slightly more lipophilic and were taken up to a greater extent by the bacteria, they were consistently less active than their linear counterparts. Of particular note, the cellular accumulation of the oligonuclear ruthenium complexes was greater in the Gram negative strains compared to that in the Gram positive S. aureus and MRSA. The results demonstrate that the lower antimicrobial activity of polypyridylruthenium(II) complexes towards Gram negative bacteria, particularly P. aeruginosa, is not strongly correlated to the cellular accumulation but rather to a lower intrinsic ability to kill the Gram negative cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284467
2-Methoxyethanol, anhydrous, 99.8%
C3H8O2
270482
2-Methoxyethanol, for HPLC, ≥99.9%
C3H8O2
360503
2-Methoxyethanol, contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%
C3H8O2
185469
2-Methoxyethanol, ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer
C3H8O2
64719
2-Methoxyethanol, for amino acid analysis, ≥99.5% (GC)
C3H8O2
88907
2-Methoxyethanol, analytical standard
C3H8O2
33457
2-Methoxyethanol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
C3H8O2
PHR1544
2-Methoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O2
216593
Ammonium hexafluorophosphate, 99.99% trace metals basis
H4F6NP
201138
Ammonium hexafluorophosphate, ≥95%
H4F6NP
A9393
Ampicillin, anhydrous, 96.0-100.5% (anhydrous basis)
C16H19N3O4S
59349
Ampicillin, analytical standard
C16H19N3O4S
A1593
Ampicillin, meets USP testing specifications
C16H19N3O4S · 3H2O
A1000000
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
C16H19N3O4S
1033000
Ampicillin, United States Pharmacopeia (USP) Reference Standard
C16H19N3O4S
746460
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
ClLi
793620
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
ClLi
L4408
Lithium chloride, BioXtra, ≥99.0% (titration)
ClLi
62476
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
ClLi
L9650
Lithium chloride, for molecular biology, ≥99%
ClLi
429457
Lithium chloride, anhydrous, beads, −10 mesh, 99.998% trace metals basis
ClLi
449881
Lithium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
ClLi
203637
Lithium chloride, powder, ≥99.99% trace metals basis
ClLi
HT-LICLSLUG
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
751898
Lithium chloride, anhydrous, chunks, 99.99% trace metals basis
ClLi
L7026
Lithium chloride solution, 8 M, for molecular biology, ≥99%
ClLi
694576
Lithium chloride solution, 1 M in 1,3-dimethyl-2-imidazolidinone
ClLi
62483
Lithium chloride solution, 1 M in glacial acetic acid
ClLi
62481
Lithium chloride solution, 1 M in ethanol
ClLi
667544
Lithium chloride solution, 0.5 M in anhydrous tetrahydrofuran
ClLi