EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
Simultaneous analysis of vitamins B in pharmaceuticals and dietary supplements by capillary electrophoresis hyphenated with triple quadrupole mass spectrometry.
PMID 25272936
Abstract
The advanced hyphenated method based on capillary electrophoresis (CE) on-line combined with electrospray ionization (ESI) and tandem mass spectrometry (MS/MS, here triple quadrupole, QqQ) was developed and applied in pharmaceutical analysis in this work. Under the optimized CE separation (background electrolyte: 50 mmol/L formic acid with pH 2.05), ESI ionization (sheath liquid: 0.1% v/v formic acid in 50% v/v aq methanol) and QqQ detection (100-160V fragmentor voltage, 15-25eV collision energy, depending on the type of analyte) conditions, highly efficient, reproducible, accurate, sensitive (sub microg/mL LODs) and selective (each analyte was unequivocally specified by two characteristic MS signals, i.e. qualifier, quantifier) determination of vitamins B was achieved. All vitamins B, namely thiamine, riboflavin, nicotinamide, nicotinic acid, pantothenic acid, pyridoxine, biotin, folic acid and cyanocobalamine, were simultaneously determined in various dosage forms (injection solution, tablets, and effervescent tablets) within ca. 16 min. The main benefits of the proposed CE-ESI-QqQ method in comparison with the CE-UV ones are considerably enhanced sensitivity and selectivity. Other benefits are low cost, simplicity, flexibility and environmental aspects when comparing CE-ESI-QqQ with HPLC-MS. Successful validation and application of the proposed CE-ESI-QqQ method suggest its routine use in highly effective and reliable multi-drug quality control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1232
Calcium-d-pantothenate, Pharmaceutical Secondary Standard; Certified Reference Material
C9H16NO5 · 0.5Ca
89126
Cetirizin dihydrochloride, ≥98.0% (HPLC)
C21H25ClN2O3 · 2HCl
C3618
Cetirizine dihydrochloride, ≥98% (HPLC)
C21H25ClN2O3 · 2HCl
C0980650
Cetirizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
C21H25ClN2O3 · 2HCl
1102929
Cetirizine dihydrochloride, United States Pharmacopeia (USP) Reference Standard
C21H25ClN2O3 · 2HCl
Y0001197
Cetirizine for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H25ClN2O3 · 2HCl
1461003
Niacin, United States Pharmacopeia (USP) Reference Standard
C6H5NO2
PHR1276
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
C6H5NO2
N4126
Nicotinic acid, ≥98%
C6H5NO2
72309
Nicotinic acid, ≥99.5% (HPLC)
C6H5NO2
N0761
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
C6H5NO2
47864
Nicotinic acid, analytical standard
C6H5NO2
N5410
Nicotinic acid, meets USP testing specifications
C6H5NO2
N0700000
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard
C6H5NO2
P9755
Pyridoxine hydrochloride, ≥98% (HPLC)
C8H11NO3 · HCl
P4722
Pyridoxine hydrochloride, meets USP testing specifications
C8H11NO3 · HCl
PHR1036
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C8H11NO3 · HCl
P4100000
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C8H11NO3 · HCl
1587001
Pyridoxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C8H11NO3 · HCl
P6280
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C8H11NO3 · HCl
47862
Pyridoxine Hydrochloride (B6), analytical standard
C8H11NO3 · HCl
PHR1054
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
C17H20N4O6
1603006
Riboflavin, United States Pharmacopeia (USP) Reference Standard
C17H20N4O6
R0600000
Riboflavin, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
Y0000757
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
T4625
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
C12H17ClN4OS · HCl
T1270
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C12H17ClN4OS · HCl
51622
Thiamine hydrochloride, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C12H17ClN4OS · HCl
PHR1037
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C12H17ClN4OS · HCl
1656002
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C12H17ClN4OS · HCl