EMAIL THIS PAGE TO A FRIEND
Apoptosis : an international journal on programmed cell death
Radiation-induced glioblastoma signaling cascade regulates viability, apoptosis and differentiation of neural stem cells (NSC).
PMID 25273222
Abstract
Ionizing radiation alone or in combination with chemotherapy is the main treatment modality for brain tumors including glioblastoma. Adult neurons and astrocytes demonstrate substantial radioresistance; in contrast, human neural stem cells (NSC) are highly sensitive to radiation via induction of apoptosis. Irradiation of tumor cells has the potential risk of affecting the viability and function of NSC. In this study, we have evaluated the effects of irradiated glioblastoma cells on viability, proliferation and differentiation potential of non-irradiated (bystander) NSC through radiation-induced signaling cascades. Using media transfer experiments, we demonstrated significant effects of the U87MG glioblastoma secretome after gamma-irradiation on apoptosis in non-irradiated NSC. Addition of anti-TRAIL antibody to the transferred media partially suppressed apoptosis in NSC. Furthermore, we observed a dramatic increase in the production and secretion of IL8, TGFβ1 and IL6 by irradiated glioblastoma cells, which could promote glioblastoma cell survival and modify the effects of death factors in bystander NSC. While differentiation of NSC into neurons and astrocytes occurred efficiently with the corresponding differentiation media, pretreatment of NSC for 8 h with medium from irradiated glioblastoma cells selectively suppressed the differentiation of NSC into neurons, but not into astrocytes. Exogenous IL8 and TGFβ1 increased NSC/NPC survival, but also suppressed neuronal differentiation. On the other hand, IL6 was known to positively affect survival and differentiation of astrocyte progenitors. We established a U87MG neurosphere culture that was substantially enriched by SOX2(+) and CD133(+) glioma stem-like cells (GSC). Gamma-irradiation up-regulated apoptotic death in GSC via the FasL/Fas pathway. Media transfer experiments from irradiated GSC to non-targeted NSC again demonstrated induction of apoptosis and suppression of neuronal differentiation of NSC. In summary, intercellular communication between glioblastoma cells and bystander NSC/NPC could be involved in the amplification of cancer pathology in the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4502549
Anti-BAX antibody produced in rabbit, affinity isolated antibody
HPA027878
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
P7874
Anti-p53 antibody, Mouse monoclonal, clone DO-2, purified from hybridoma cell culture
SAB4300033
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
SAB4504541
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody
S2690
Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701800
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S9072 Anti-SOX2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4300669
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB3500363
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701973
Anti-SOX2 antibody produced in rabbit
SAB2701974
Anti-SOX2 antibody produced in rabbit
S1451
Anti-Sox2 antibody, Mouse monoclonal, clone SOX2-6, purified from hybridoma cell culture
SAB2103958
Anti-SOX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300708
Anti-STAT3 (Ab-705) antibody produced in rabbit, affinity isolated antibody
SAB4300327
Anti-STAT3 (Ab-727) antibody produced in rabbit, affinity isolated antibody
SAB2500993
Anti-STAT3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104461
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2104912
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
SAB2102317
Anti-STAT3 antibody produced in rabbit, affinity isolated antibody
S5933 Anti-STAT3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody