EMAIL THIS PAGE TO A FRIEND
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging
Synthesis and preclinical evaluation of [¹⁸F]FCHC for neuroimaging of fatty acid amide hydrolase.
PMID 25273322
Abstract
Fatty acid amide hydrolase (FAAH), a catabolic enzyme which regulates lipid transmitters in the endocannabinoid system, is an avidly sought therapeutic and positron emission tomography (PET) imaging target for studies involving addiction and neurological disorders. We report the synthesis of a new fluorine-18-labeled FAAH inhibitor, trans-3-(4, 5-dihydrooxazol-2-yl)phenyl-4-[(18)F]fluorocyclohexylcarbamate ([(18)F]FCHC), and its evaluation in rat brain. The synthesis of [(18)F]FCHC was conducted via a 3-step, 1-pot reaction, resulting in uncorrected radiochemical yields between 10 and 20% (n = 5) relative to [(18)F]fluoride, with specific activities of >5 Ci/μmol at the end of the synthesis. The radiosynthesis was seamlessly automated using a commercial radiofluorination apparatus. Ex vivo biodistribution and preliminary PET imaging studies were carried out in male Sprague-Dawley rats. Rat brain biodistribution at 2 min post-injection showed a standard uptake value of 4.6 ± 0.1 in the cortex, which increased to 7.8 ± 0.1 at 40 min. Pretreatment with the selective FAAH inhibitor URB597 reduced uptake of radioactivity in all brain regions by >90%, with 98 % blockade in the FAAH-rich cortex. PET imaging was consistent with biodistribution studies. [(18)F]FCHC appears to be a highly sensitive (18)F-labeled radiotracer for imaging FAAH in the central nervous system, and these results warrant further imaging in nonhuman primates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151858
Chloroform-d, "100%", 99.96 atom % D
CCl3D
494275
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
CCl3D
431915
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
CCl3D
151823
Chloroform-d, 99.8 atom % D
CCl3D
225789
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
CCl3D
612200
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
CCl3D
434876
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
CCl3D
151831
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
CCl3D
416754
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
CCl3D
530735
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
CCl3D
570699
Chloroform-d, ≥99.8 atom % D, anhydrous
CCl3D
156914
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
453323
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
424005
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
417939
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
417920
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 1 % (v/v) TMS
C2D6OS
570672
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
C2D6OS
569585
Dimethyl sulfoxide-d6, 99.9 atom % D, anhydrous
C2D6OS
151874
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
522058
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
612324
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
C2D6OS
716731
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
C2D6OS
296147
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
574341
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
185965
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
C2D6OS
175943
Dimethyl sulfoxide-d6, 99.5 atom % D
C2D6OS
724335
Dimethyl sulfoxide-d6, contains 0.07 wt. % TSP-d4, 99.9 atom % D, 99% (CP)
C2D6OS
U4133
URB597, ≥98% (HPLC), powder
C20H22N2O3