EMAIL THIS PAGE TO A FRIEND
Parasitology Research
Triosephosphate isomerase of Taenia solium (TTPI): phage display and antibodies as tools for finding target regions to inhibit catalytic activity.
PMID 25273631