EMAIL THIS PAGE TO A FRIEND
Journal of Human Hypertension
Assessment of toxic elements in the samples of different cigarettes and their effect on the essential elemental status in the biological samples of Irish hypertensive consumers.
PMID 25273861