EMAIL THIS PAGE TO A FRIEND
Journal of Virology
Nipah virion entry kinetics, composition, and conformational changes determined by enzymatic virus-like particles and new flow virometry tools.
PMID 25275126