EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
New developments of RNAi in Paracoccidioides brasiliensis: prospects for high-throughput, genome-wide, functional genomics.
PMID 25275433
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D134406
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
C10H12O4