EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Preparation, characteristics and assessment of a novel gelatin-chitosan sponge scaffold as skin tissue engineering material.
PMID 25275938