EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Isophoronediamine (IPD) dermatitis in Spain.
PMID 2527720
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
118184
5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans, ≥99%
C10H22N2