EMAIL THIS PAGE TO A FRIEND
Nature communications
An endocytosis pathway initiated through neuropilin-1 and regulated by nutrient availability.
PMID 25277522
Abstract
Neuropilins (NRPs) are trans-membrane receptors involved in axon guidance and vascular development. Many growth factors and other signalling molecules bind to NRPs through a carboxy (C)-terminal, basic sequence motif (C-end Rule or CendR motif). Peptides with this motif (CendR peptides) are taken up into cells by endocytosis. Tumour-homing CendR peptides penetrate through tumour tissue and have shown utility in enhancing drug delivery into tumours. Here we show, using RNAi screening and subsequent validation studies, that NRP1-mediated endocytosis of CendR peptides is distinct from known endocytic pathways. Ultrastructurally, CendR endocytosis resembles macropinocytosis, but is mechanistically different. We also show that nutrient-sensing networks such as mTOR signalling regulate CendR endocytosis and subsequent intercellular transport of CendR cargo, both of which are stimulated by nutrient depletion. As CendR is a bulk transport pathway, our results suggest a role for it in nutrient transport; CendR-enhanced drug delivery then makes use of this natural pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
324558
Ethylene glycol, anhydrous, 99.8%
C2H6O2
03747
Ethylene glycol, BioUltra, ≥99.5% (GC)
C2H6O2
85978
Ethylene glycol, analytical standard
C2H6O2
1265515
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
C2H6O2
PHR1046
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O2
24204
Ethylene glycol, puriss., ≥99.5% (GC)
C2H6O2
61941 Ethylene glycol 5 M solution
C2H6O2
721980 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
C2H6O2
721972 Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
C2H6O2
W326305
L-Cysteine, ≥97%, FG
C3H7NO2S
168149
L-Cysteine, 97%
C3H7NO2S
C7352
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
C3H7NO2S
C5360
L-Cysteine, PharmaGrade, Ajinomoto, from non-animal source, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S
30089
L-Cysteine, BioUltra, ≥98.5% (RT)
C3H7NO2S
778672
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
C3H7NO2S
95437
L-Cysteine, certified reference material, TraceCERT®
C3H7NO2S
EHU095021 MISSION® esiRNA, esiRNA human NELL1 (esiRNA1)
EHU134551 MISSION® esiRNA, esiRNA human NRP1 (esiRNA1)
EMU201661 MISSION® esiRNA, esiRNA targeting mouse Nell1
M1177
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
C21H39NO6
37094
Rapamycin, VETRANAL, analytical standard
C51H79NO13
S8157
Silver nitrate, BioXtra, >99% (titration)
AgNO3
S7276
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
AgNO3
204390
Silver nitrate, 99.9999% trace metals basis
AgNO3
85229
Silver nitrate, tested according to Ph.Eur.
AgNO3
792276
Silver nitrate, anhydrous, ≥99.999% trace metals basis
AgNO3
248762
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
AgNO3
85193
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
AgNO3
11691
Silver standard for ICP, for ICP, ready-to-use, in nitric acid
AgNO3
1613655
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
CHNaO3