EMAIL THIS PAGE TO A FRIEND
Skeletal radiology
Labral and cartilage abnormalities in young patients with hip pain: accuracy of 3-Tesla indirect MR arthrography.
PMID 25277527
Abstract
Assess the diagnostic accuracy of 3-T indirect magnetic resonance arthrography (iMRA) for hip cartilage and labral pathology detection using arthroscopy as the reference standard and compare it to the published performance of direct magnetic resonance arthrography (dMRA). Between 2009 and 2011, 290 patients suspected of having femoroacetabular impingement underwent iMRA. Our study group consisted of 41 of these patients (17 males, mean age 35xa0years; 24 females, mean age 33xa0years) who did not have a prior history of hip surgery and who subsequently underwent arthroscopy. Two experienced musculoskeletal radiologists separately evaluated the randomized and anonymized studies for the presence and quadrant location of labral and cartilage pathology. These recorded data were compared to arthroscopic reports. Forty-one patients had labral pathology, 34 patients had acetabular and 5 patients had femoral cartilage pathology at arthroscopy. Sensitivity, specificity, accuracy, negative- and positive-predictive values for labral lesion detection were respectively 98, 99, 99, 99 and 98xa0%; for acetabular cartilage lesion detection they were 69, 98, 89, 87 and 95xa0%; for femoral cartilage lesion detection they were 69, 95, 93 and 39xa0%. Sensitivities of iMRA by quadrant (anteroinferior, anterosuperior, posteroinferior, posterosuperior) for the labrum were 100.0, 95.0, NA and 85.7xa0%, for acetabular cartilage were NA, 58.8, NA and 39.5xa0% and for femoral cartilage were 50.0, 33.3, 75.0 and 75.0xa0%). NA indicates results not available because of the absence of findings in those quadrants. Specificities of iMRA by quadrant (anteroinferior, anterosuperior, posteroinferior, posterosuperior) for the labrum (95.0, 100.0, 95.1, 67.5xa0%), acetabular (100.0, 85.7, 92.6, 79.5xa0%) and femoral cartilage (100.0, 94.7, 96.2, 85.9xa0%). iMRA at 3xa0T is accurate in detecting labral pathology suggesting that it is a viable alternative to dMRA.