EMAIL THIS PAGE TO A FRIEND
Clinica Chimica Acta; International Journal of Clinical Chemistry
Simple and robust ICP-MS method for simultaneous determination of serum Co and Cr in routine clinical practice.
PMID 25278349