EMAIL THIS PAGE TO A FRIEND
International Journal of Immunopathology and Pharmacology
Human mesenchymal stem cells produce bioactive neurotrophic factors: source, individual variability and differentiation issues.
PMID 25280030