EMAIL THIS PAGE TO A FRIEND
Vaccine
Vaccination with liposomal poly(I:C) induces discordant maturation of migratory dendritic cell subsets and anti-viral gene signatures in afferent lymph cells.
PMID 25280435