EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Construction of a quadruple auxotrophic mutant of an industrial polyploid saccharomyces cerevisiae strain by using RNA-guided Cas9 nuclease.
PMID 25281382