EMAIL THIS PAGE TO A FRIEND
Sudebno-meditsinskaia ekspertiza
[Determination of zimarin in urine].
PMID 2528227
Abstract
Private technique of extraction isolation and purification, chromatographic detection and photometric determination of zimarin in urine is suggested. Detection limit is 0.01 mg, determination limit is 0.1 mg of glycoside in 100 ml of urine. Method makes it possible to detect 66-80% of zimarin added to 100 ml of urine in quantities 0.5-0.1 mg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30030
Cymarin, ≥96% (HPLC)
C30H44O9