EMAIL THIS PAGE TO A FRIEND
Journal of Hazardous Materials
Long-term trends of PBDEs, triclosan, and triclocarban in biosolids from a wastewater treatment plant in the Mid-Atlantic region of the US.
PMID 25282513